Sterilin™ 100mm Square Petri Dishes
Sterilin™ 100mm Square Petri Dishes
Actual product may vary
Fisherbrand
Sterilin™ 100mm Square Petri Dishes
Conduct various applications with Thermo ScientificTM SterilinTM 100mm Square Petri Dishes, available in both compartmentalized and non-compartmentalized formats.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell CountVent
FKS113492731No
FKS1133927325Selectable
Catalog number FKS11349273
Price (SGD)
1,238.00
PK
Add to cart
Well Count:
1
Vent:
No
Price (SGD)
1,238.00
PK
Add to cart
Conduct various applications with Thermo Scientific™ Sterilin™ 100mm Square Petri Dishes, available in both compartmentalized and non-compartmentalized formats. Manufactured in Clean Room Class 7 Conditions to exclude microbiological contamination.
NOT AVAILABLE IN AMERICA
Non-compartmentalized:
  • Ideal for antibiotic sensitivity testing when a large surface area and very flat base is required
Compartmentalized:
  • Ideal for small volume liquid media work or for sample storage
  • 25 compartments each have a surface area of 3.7cm2 and a capacity of 5mL
  • Lid features selectable venting or non-venting positions where the lid turns through 90° to allow for venting or non-venting of the dish
Ordering Information:
Not currently available in North America
Specifications
DescriptionPetri dish, 100mm
Bottom ShapeFlat
Capacity (Metric)5 mL (Single Compartment)
ShapeSquare
SterilityNon-sterile
TypePetri Dish
Well Count1
No. per Case120
No. per Pack4
VentNo
Unit SizePK