Sterilin™ Cones
Sterilin™ Cones
Actual product may vary
Fisherbrand
Sterilin™ Cones
Safely mount your filter funnels without risk of chipping or slipping. Made of natural rubber, cones are also useful for mounting crucibles onto adapters.
Have Questions?
Catalog NumberDescription
FKS11349183Set of 6 cones; Assorted
Catalog number FKS11349183
Price (SGD)
197.00
EA
Add to cart
Description:
Set of 6 cones; Assorted
Price (SGD)
197.00
EA
Add to cart
Safely mount your filter funnels without risk of chipping or slipping. Thermo Scientific™ Sterilin™ Cones are made of natural rubber, and are also useful for mounting crucibles onto adapters.
  • Set of six cones
  • Manufactured from natural rubber
  • For mounting filter funnels into Büchner flasks
  • Also suitable for mounting Gooch crucibles onto adapters
Ordering Information:
  • Not available in all geographic regions
Specifications
DescriptionSet of 6 cones; Assorted
MaterialRubber
For Use With (Equipment)Büchner flasks and Gooch crucibles
IncludesSet of six cones
Unit SizeEA