LDPE Narrow Mouth Bottles with EPE-lined Black Polyethylene Screw Cap
LDPE Narrow Mouth Bottles with EPE-lined Black Polyethylene Screw Cap
Actual product may vary
Fisher Catalogue
LDPE Narrow Mouth Bottles with EPE-lined Black Polyethylene Screw Cap
View and identify colored liquids quickly and easily with translucent bottles. Azpack™ LDPE Narrow Mouth Bottles with EPE-lined Black Polyethylene Screw Cap are highly resistant to dilute and concentrated acids, alcohols, bases, and esters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Inner Neck
FKS11348043500 mL18 mm
FKS11338043250 mL15 mm
Catalog number FKS11348043
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Diameter (Metric) Inner Neck:
18 mm
  • Translucent
  • Lightweight LDPE
  • Polyethylene screw cap with EPE Liner
  • Supplied in packs of 10
Specifications
Capacity (Metric)500 mL
ColorTranslucent
Diameter (Metric) Inner Neck18 mm
MaterialLDPE
Quantity10/PK
Closure ColorBlack
Closure IncludedYes
MouthNarrow
Height (Metric)172 mm
Closure LinerEPE
Closure MaterialPolyethylene
Diameter (Metric)70 mm
Closure TypeScrew Cap
Unit SizePK