Whatman™ Grade 602H Qualitative Filter Papers
Whatman™ Grade 602H Qualitative Filter Papers
Actual product may vary
Fisherbrand
Whatman™ Grade 602H Qualitative Filter Papers
Collects very small particles and removes fine precipitates. Whatman™ Grade 602H Qualitative Filter Papers are used in the beverage industry to determine residual sugar, acidic spectra, refractometric analysis and HPLC.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS11345244150 mm
FIS09-924-303150 mm
Catalog number FKS11345244
Price (SGD)
196.00
PK
Add to cart
Diameter (Metric):
150 mm
Price (SGD)
196.00
PK
Add to cart
Collects very small particles and removes fine precipitates. Whatman™ Grade 602H Qualitative Filter Papers are used in the beverage industry to determine residual sugar, acidic spectra, refractometric analysis and HPLC.
Cellulose filters used with qualitative analytical techniques to determine and identify materials
  • Residual sugar determination
  • Acidic spectra
  • Refractometric analysis
  • HPLC
Specifications
DescriptionGrade 602 h Qualitative Filter Paper Standard Grade, circle, 150 mm
FormatDisc
MaterialCellulose
Particle Retention<2 μm
Filtration SpeedApprox. 375 s
Flow RateSlow
For Use With (Application)A standard grade filter esed in sample preparation, e.g., in the beverage industry for residual sugar determination, acidic spectra, refractometric analysis, and HPLC.
GradeGrade 602 h
Quantity100 Pk.
Thickness (Metric)160 μm
Diameter (Metric)150 mm
Unit SizePK