Isothermal Ice Pan
Isothermal Ice Pan
Actual product may vary
Fisher Catalogue
Isothermal Ice Pan
Provides high durability and excellent insulation. IsoThermal Ice Pans are the ideal cooling containers for ultra-cold uses, including ice, ice slurries and CO2 pellets. Convenient and easy to use, with lids that nest securely under the bucket. Available in three color-coded sizes.
Have Questions?
Catalog NumberCapacity (Metric)ColorDimensions (L x W x H)
FKS113440859 LRed52.1 x 29.2 x 16.3 cm
Catalog number FKS11344085
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
9 L
Color:
Red
Dimensions (L x W x H):
52.1 x 29.2 x 16.3 cm
Ideal containers for ultra-cold environments.
  • Rigid single-molded polyurethane provides high durability and stability when chilling samples.
  • Excellent insulation properties
  • The close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob.
  • Containers will not sweat or leak and will not break or deform with normal use.
  • Withstands temperatures from -80°C to +95°C.
Convenient, user-friendly design:
  • Lids nest conveniently under the bucket because they are keyed to fit together.
  • Molded handles ensure safe handling and transport.
  • Precise pour spout for easy emptying.
  • Tapered design for easy stacking.
  • Color-coded (green, blue, red) for easy organization and identification.
Specifications
Height (Metric)16.3 cm
Height (English)6.4 in.
Length (Metric)52.1 cm
Length (English)2.1 in.
Width (Metric)29.2 cm
Width (English)11.5 in.
Dimensions (L x W x H)52.1 x 29.2 x 16.3 cm
Capacity (English)2.4 gal.
For Use With (Equipment)Dry ice, ice water, ice-salt slurries, etc.
MaterialPolyurethane
TypeIce Pan
ColorRed
Capacity (Metric)9 L
AutoclavableNot Autoclavable
Temperature ControlRefrigerated
Temperature Range-80°C to +95°C
Unit SizePK