Azlon™ Polypropylene Cylinders with Printed Graduations
Azlon™ Polypropylene Cylinders with Printed Graduations
Actual product may vary
Azlon™ Polypropylene Cylinders with Printed Graduations
Printed with pin sharp blue graduations for enhanced visibility. Azlon™ Polypropylene Cylinders with Printed Graduations provide a reliable solution for measuring the volume of liquids in a laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS113435942000 mL20 mL
FKS113335941000 mL10 mL
Catalog number FKS11343594
Price (SGD)
93.00
EA
Add to cart
Capacity (Metric):
2000 mL
Graduations:
20 mL
Price (SGD)
93.00
EA
Add to cart
Printed with pin sharp blue graduations for enhanced visibility. Azlon™ Polypropylene Cylinders with Printed Graduations provide a reliable solution for measuring the volume of liquids in a laboratory.
  • Hexagonal base improves stability
  • Anti-suction feature prevents cylinder from sticking to wet surfaces and minimizes spillage
  • Ultra clear polypropylene provides enhanced visibility and chemical resistance
  • Class B
  • Conforms to ISO 6706
Specifications
Capacity (Metric)2000 mL
MaterialPolypropylene
Tolerance±20 mL
Certifications/ComplianceISO 6706
ClassB
ScaleBlue
Diameter (Metric)83 mm
Quantity1PK
Graduations20 mL
Scale ColorBlue
Base TypeHexagonal
Height (English)21.06 in.
Height (Metric)535 mm
TypeUngraduated Cylinder
Unit SizeEA