1x1 image pixel for data collection
Whatman™ Qualitative Filter Paper: Grade 113V Circles
Whatman™ Qualitative Filter Paper: Grade 113V Circles
Actual product may vary
Fisherbrand
Whatman™ Qualitative Filter Paper: Grade 113V Circles
Saves time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses. Whatman™ Qualitative Filter Paper: Grade 113V Circles boast the fastest flow rate of the qualitative grades. Crepe surface promotes ultra high loading capacity.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11342925150 mm
Catalog number FKS11342925
Price (SGD)
360.00
PK
Add to cart
Diameter (Metric):
150 mm
Price (SGD)
360.00
PK
Add to cart
Saves time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses. Whatman™ Qualitative Filter Paper: Grade 113V Circles boast the fastest flow rate of the qualitative grades. Crepe surface promotes ultra high loading capacity.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate.
  • Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous 
  • Fastest flow rate of the qualitative grades
  • Crepe surface
  • Thickest filter paper in the qualitative range
  • Retention of coarse particles: nominal particle retention rating of 30 μm
  • Pleated for easy use in applications involving a conical filter funnel
Note: Resins contain nitrogen so these grades should not be used in Kjeldahl estimations or similar operations.
Specifications
DescriptionGrade 113V Creped Filter Paper, Wet-Strengthened, 150 mm circle (100 pcs)
FormatPleated Circle
MaterialCellulose
Particle Retention30 μm
Pore Size>30 μm
Filtration SpeedHerzberg 28 sec.
Flow RateFast
For Use With (Application)GE’s Whatman Grade 113V creped filter paper, for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
GradeGrade 113V
Quantity100/Pk.
Thickness (Metric)420 μm
TypeQualitative Filter Paper
Diameter (English)5.9 in.
Diameter (Metric)150 mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link