Whatman™ Qualitative Filter Paper: Grade 2 Circles
Whatman™ Qualitative Filter Paper: Grade 2 Circles
Actual product may vary
Fisherbrand
Whatman™ Qualitative Filter Paper: Grade 2 Circles
Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1134282542.5 mm
FIS09-810B55 mm
Catalog number FKS11342825
Price (SGD)
54.00
PK
Add to cart
Diameter (Metric):
42.5 mm
Price (SGD)
54.00
PK
Add to cart
Retains and absorbs more than Grade 1 filters, with a corresponding increase in filtration time. Whatman™ Qualitative Filter Paper: Grade 2 Circles are used for monitoring specific contaminants in the atmosphere and in soil testing.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids. 
  • Particle Retention: 8μm
  • Porosity: Medium fine
  • Filtration speed: ASTM™, 55 sec.; Herzberg, 240 sec.
  • Surface: Smooth
  • Holds soil nutrient in plant growth trials
  • Used for soil analysis and seed testing procedures in the agricultural industry
Specifications
DescriptionGrade 2 Qualitative Filter Paper Standard Grade, circle, 42.5 mm
FormatDisc
MaterialCellulose
Particle Retention>8 μm
Filtration Speed240 sec.
Flow RateMedium
For Use With (Application)A standard grade filter used for monitoring specific contaminants in the atmosphere and in soil testing.
GradeGrade 2
PorosityMedium Fine
Quantity100 Pk.
Thickness (Metric)190 μm
TypeQualitative Filter Paper
Diameter (English)1.67 in.
Diameter (Metric)42.5 mm
Unit SizePK