SP Scienceware™ LabMat™ Bench Liners
SP Scienceware™ LabMat™ Bench Liners
Actual product may vary
SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)ColorDescription
FKS1134219450 ft. x 20 in.OrangeOne single roll, 50 ft.
FIS11-999-1850 ft. x 20 in.WhiteOne single roll, 50 ft.
Catalog number FKS11342194
Price (SGD)
390.00
ROL
Add to cart
Dimensions (L x W):
50 ft. x 20 in.
Color:
Orange
Description:
One single roll, 50 ft.
Price (SGD)
390.00
ROL
Add to cart
  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials
Specifications
Product TypeBench Protector
MaterialPolyethylene
Absorbancy400 mL/m2
Length (English)50 ft.
Length (Metric)15.2 m
Width (English)20 in.
Width (Metric)508 mm
Dimensions (L x W)50 ft. x 20 in.
ColorOrange
DescriptionOne single roll, 50 ft.
DisposableSingle-use
For Use With (Application)Simplify cleanup and safeguard work surfaces from stains, spills and wear
QuantityEach
Unit SizeROL