Add detachable tubing to a glass vescle with a glass tubing adapter. QuickFit™ straight tubing adapters come in different sizes to fit containers or varying sizes.
Have Questions?
Catalog Number
Standard Taper Joint No.
FKS11342045
19/26
Catalog number FKS11342045
Price (SGD)
140.00
EA
Add to cart
Standard Taper Joint No.:
19/26
Price (SGD)
140.00
EA
Add to cart
With straight connection for the attachment of rubber hose etc
Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
For Use With (Equipment)Flexible tubing, Rubber hose