Plastilab™ Finned Plug for Disposable Tubes
Plastilab™ Finned Plug for Disposable Tubes
Actual product may vary
Fisherbrand
Plastilab™ Finned Plug for Disposable Tubes
Designed with flexible fins to create a snug fit.
Have Questions?
Catalog NumberColor
FKS11341195Yellow
Catalog number FKS11341195
Price (SGD)
98.00
PK
Add to cart
Color:
Yellow
Price (SGD)
98.00
PK
Add to cart
Designed with flexible fins to create a snug fit.
  • Fits tubes 15 to 17mm 
  • Available in a variety of colors for coding
Specifications
Certifications/ComplianceCE-IVD
Product TypeFinned Plug
MaterialPolyethylene
ColorYellow
For Use With (Equipment)15 to 17 mm Tubes
Unit SizePK