HDPE, Heavy Duty Aspirator Bottles
HDPE, Heavy Duty Aspirator Bottles
Actual product may vary
Fisherbrand
HDPE, Heavy Duty Aspirator Bottles
For Use With: Designed for life time service, suitable for food stuffs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1134114550 L
FKS1027399110 L
FKS1050562125 L
Catalog number FKS11341145
Price (SGD)
365.00
EA
Add to cart
Capacity (Metric):
50 L
Price (SGD)
365.00
EA
Add to cart
For Use With: Designed for life time service, suitable for food stuffs
  • Designed with thick (2mm), translucent walls 
  • Includes carry handles and cap
  • Supplied with 3/4 in. BSP tap that mates with a 12mm thread in the aspirator for leakproof performance
  • Suitable for foodstuff
  • Recommended: use petroleum jelly on the thread before assembly
Image shows (left to right): 25L, 5L, 10L bottles.
Specifications
Capacity (English)13 gal.
Capacity (Metric)50 L
Diameter (Metric) Inner Neck79.5 mm
Diameter (Metric) Outer350 mm
For Use With (Application)Designed for life time service, suitable for food stuffs
Height (Metric) With Closure700 mm
IncludesClosure, Carrying Handles, Spigot
MaterialHDPE
Mouth StyleNarrow
No. per Pack2 Pk.
Product TypeAspirator Bottle
Unit SizeEA