Unex™ Tubing Clips
Unex™ Tubing Clips
Actual product may vary
Fisherbrand
Unex™ Tubing Clips
Twist nut to constrict and release tubing. Azpack™ Stainless Steel Tubing Clips are designed to distribute and apply pressure evenly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1133840310 to 14 mm
FKS1130810313 to 17 mm
FKS1132840318 to 22 mm
FKS1133810328 to 33 mm
Catalog number FKS11338403
Price (SGD)
-
Diameter (Metric):
10 to 14 mm
  • Stainless steel
  • Double-banded ring for even pressure
  • Nut and bolt included to apply proper compression
Specifications
TypeTubing Clamp
MaterialStainless Steel (H10 304, A2)
IncludesTightening bolt
Diameter (Metric)10 to 14 mm
Jaw TypeDouble-banded Ring
Unit SizeEA