Check Order Status
Tube Cleaner Brush
Actual product may vary
Fisherbrand
Tube Cleaner Brush
Cleans small bore tubes, stopcocks, and pipettes. Azpack™ Tube Cleaner Brush is available in a range of styles to meet a variety of cleaning needs.
Have Questions?
Change view
Catalog Number
Bristle Material
Diameter (Metric) Bristle
Color
FKS11338083
Extra-absorbent Fleece
6 mm
White
FKS11318083
Standard Stiffer Wire Yarn
3 mm
White
2 Options
Catalog number
FKS11338083
Price (SGD)
61.00
EA
Add to cart
Bristle Material:
Extra-absorbent Fleece
Diameter (Metric) Bristle:
6 mm
Color:
White
Price (SGD)
61.00
EA
Add to cart
To increase diameter simply double over and twist
Length: 10m
Specifications
Product Type
Pipe Cleaner
Bristle Material
Extra-absorbent Fleece
Handle Material
Flexi-stem Coil
Color
White
Diameter (Metric) Bristle
6 mm
Length (English)
393.7 in.
Length (Metric)
10 m