Isotemp™ Cooling/Heating Recirculating Chiller, 230V 50Hz
Isotemp™ Cooling/Heating Recirculating Chiller, 230V 50Hz
Actual product may vary
Fisherbrand
Isotemp™ Cooling/Heating Recirculating Chiller, 230V 50Hz
Designed with a focus on high performance, small footprint and quiet operation.
Have Questions?
Catalog NumberModelPump Type
FKS11336661500LCSUForce and Suction
Catalog number FKS11336661
Price (SGD)
19,183.00
EA
Add to cart
Model:
500LCSU
Pump Type:
Force and Suction
Price (SGD)
19,183.00
EA
Add to cart
  • Full range heating and cooling from -10°C to +80°C
  • Small reservoir ensures fast temperature response time
  • Small footprint
  • Quiet operation
Specifications
Capacity (Metric)2.8 L
Cooling CapacityAt +20°C
Pump TypeForce and Suction
Temperature Range-10°C to +80°C
Heating Capacity2.0 kW
Model500LCSU
Pump Capacity21 L/min. (805 mbar)
Temperature Stability±0.1°C
USB ConnectionsYes
Dimensions (L x W x H)419.1 x 228.6 x 635 mm
Shipping Weight (Metric)29.9 kg
Voltage230 V
Frequency50 Hz
Unit SizeEA