Scientific Shaker Flask Platforms
Scientific Shaker Flask Platforms
Actual product may vary
Fisher Catalogue
Scientific Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Have Questions?
Catalog NumberFor Use With (Equipment)No. of PositionsLength (Metric) PlatformWidth (Metric) Platform
FKS11336004Innova 2300, Excella E5/E10, I-2500/I-2500KC4076 cm46 cm
Catalog number FKS11336004
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Innova 2300, Excella E5/E10, I-2500/I-2500KC
No. of Positions:
40
Length (Metric) Platform:
76 cm
Width (Metric) Platform:
46 cm
  • Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
  • Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Holds250 to 300mL
Product TypeDedicated Shaker Platform
MaterialAluminum
Length (English) Platform30 in.
Length (Metric) Platform76 cm
Width (English) Platform18 in.
Width (Metric) Platform46 cm
For Use With (Equipment)Innova 2300, Excella E5/E10, I-2500/I-2500KC
No. of Positions40
Unit SizePK