Millex™ Samplicity™ Filters, 0.45 μm
Millex™ Samplicity™ Filters, 0.45 μm
Actual product may vary
Fisherbrand
Millex™ Samplicity™ Filters, 0.45 μm
To be used with the Samplicity Filtration System. Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
Have Questions?
Catalog NumberMaterial (Membrane)Quantity
FKS11335982Hydrophilic PTFE96 Pk.
Catalog number FKS11335982
Price (SGD)
2,368.00
PK
Add to cart
Material (Membrane):
Hydrophilic PTFE
Quantity:
96 Pk.
Price (SGD)
2,368.00
PK
Add to cart
Overview of Millex Samplicity™ Filters for the Samplicity™ G2 System
  • Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
  • Filtration for HPLC sample prep without syringes
  • Millex Samplicity™ Filters provide integrated funnels and filters for easiest sample loading
  • Available in 0.20 μm and 0.45 μm pore sizes with hydrophilic PTFE and Durapore™ PVDF membranes
Specifications
ConnectorsEasy loading funnel inlet, Outlet tip for smooth transfer to HPLC vials
DescriptionFilter Code: LCR
DisposableSingle-use
For Use With (Equipment)Samplicity G2 Filtration System
Material (Membrane)Hydrophilic PTFE
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)Sample preparation for HPLC
Housing MaterialHigh density polyethylene (HDPE) housing
Quantity96 Pk.
TypeMillex™ Samplicity™ Filter
Volume (Metric)0.3 to 1.6 mL
Diameter (Metric) Membrane25 mm
Unit SizePK