Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
FKS11334555 | Lab Pen, Fine Line |
6310-0010 | Lab Pen, Fine Line |
Mark on glass, plastic and metal with Thermo Scientific™ Nalgene™ Black Ink Lab Markers, recommended for use with Nalgene™ Polyolefin paper, notebooks and labels. These markers have caps with a convenient packet clip. Choose from fine point or broad line options.