Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
Have Questions?
Catalog NumberFor Use With (Equipment)DescriptionPorosity
FKS11332065For model MF29/3/125 Dreschel bottleStem Length: 132 mm1
Catalog number FKS11332065
Price (SGD)
356.00
EA
Add to cart
For Use With (Equipment):
For model MF29/3/125 Dreschel bottle
Description:
Stem Length: 132 mm
Porosity:
1
Price (SGD)
356.00
EA
Add to cart
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
  • Complies with BS 2461
  • Sintered dip tube disperses gas into bubbles for greater efficiency
  • Choice of sinter porosities to achieve fine bubble size
Specifications
Certifications/ComplianceBS 2461
DescriptionStem Length: 132 mm
MaterialBorosilicate Glass
For Use With (Equipment)For model MF29/3/125 Dreschel bottle
Product TypeSintered
Porosity1
Unit SizeEA