Check Order Status
PEEK Tubing
Actual product may vary
Fisherbrand
PEEK Tubing
PEEK tubing comes in a variety of internal diameter sizes. Cytiva Lifescience™ PEEK Tubing is an important chromatography accessory.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
FKS11330012
0.75 mm
FKS11555724
0.5 mm
2 Options
Catalog number
FKS11330012
Price (SGD)
519.00
EA
Add to cart
Diameter (Metric) Inner:
0.75 mm
Price (SGD)
519.00
EA
Add to cart
Polyetheretherketone (PEEK)
A variety of sizes are available
Specifications
Type
Tubing
Description
PEEK Tubing, 2 m, i.d. 0.75 mm, o.d. 1/16 in.
Diameter (Metric) Inner
0.75 mm
Diameter (Metric) Outer
1.58 mm
Overall Length (Metric)
2 m
Material
PEEK