Whatman™ Cellulose Acetate Membranes
Whatman™ Cellulose Acetate Membranes
Actual product may vary
Fisher Catalogue
Whatman™ Cellulose Acetate Membranes
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
Have Questions?
Catalog NumberPore SizeDiameter (Metric)Membrane TypeSterility
FKS113290240.45 μm25 mmOE 67Non-sterile
Catalog number FKS11329024
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.45 μm
Diameter (Metric):
25 mm
Membrane Type:
OE 67
Sterility:
Non-sterile
Prepares liquid scintillation cocktail for scintillation counter measurements. Whatman™ Cellulose Acetate Membranes are highly heat resistant and can filter hot liquids and hot gases.
  • Low protein binding capacity
  • Hydrophilic
  • Improved solvent resistance and heat resistance
  • Can be used for liquids or gases up to 180°C
  • Sterilizable by any method without sacrificing integrity
For filtration with aqueous or alcoholic solvents and gases
Specifications
MaterialCellulose Acetate
Pore Size0.45 μm
SterilityNon-sterile
Flow Rate12s/100mL/12.5cm2
HydrophobicityHydrophilic
Membrane TypeOE 67
No. per Pack100/Pk.
Diameter (Metric)25 mm
Unit SizePK