Check Order Status
Whatman™ Glass Fiber Filter Papers - Grade 31
Actual product may vary
Fisherbrand
Whatman™ Glass Fiber Filter Papers - Grade 31
Offers fine particle retention and high flow rate, as well as good loading capacity
Have Questions?
Catalog Number
Diameter (Metric)
FKS11328904
5.5 cm
Catalog number
FKS11328904
Price (SGD)
250.00
PK
Add to cart
Diameter (Metric):
5.5 cm
Price (SGD)
250.00
PK
Add to cart
Specifications
Material
Glass Fiber
Flow Rate
Fast
For Use With (Application)
Fine particle retention and high flow rate, as well as good loading capacity
Grade
31
Diameter (English)
2.1 in.
Diameter (Metric)
5.5 cm