Single-Metric Scale Reusable Graduated Cylinders, TC with Funnel Top
Single-Metric Scale Reusable Graduated Cylinders, TC with Funnel Top
Actual product may vary
Fisherbrand
Single-Metric Scale Reusable Graduated Cylinders, TC with Funnel Top
Translucent polypropylene with raised graduations at primary scale points
Have Questions?
Catalog NumberCapacity (Metric)
FKS11327871500 mL
Catalog number FKS11327871
Price (SGD)
749.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
749.00
PK
Add to cart
Translucent polypropylene graduated cylinders are manufactured with raised graduations at the ­primary scale points. Cylinders have a hexagonal base and are calibrated 'To Contain'. Tolerances according to ISO 6706. Exposure to temperatures above 60°C may affect accuracy.
Specifications
Capacity (Metric)500 mL
MaterialPolypropylene
Tolerance±5 mL
Certifications/ComplianceISO 6706
No. per Pack1/Pk.
No. per Case6/Cs.
CalibrationTo Contain
DisposableReusable
Diameter (English)2.12 in.
ScaleSingle
Diameter (Metric)5.4 cm
Graduations5 mL
AutoclavableAutoclavable
Max. Temperature125°C
Subdivision5 mL
ShapeRound
Diameter (Metric) Inside54 mm
Diameter (English) Inside2.12 in.
SterilityNon-sterile
Base TypeHexagonal
Height (English)14.17 in.
Height (Metric)360 mm
TypeGraduated Cylinder
Unit SizePK