Spring Loaded Clamp
Spring Loaded Clamp
Actual product may vary
Fisherbrand
Spring Loaded Clamp
For use with burettes. Azlon™ Burette Clamps contain rubber grips to prevent slipping.
Have Questions?
Catalog NumberType
FKS11327794Burette Clamp (Single)
Catalog number FKS11327794
Price (SGD)
108.00
EA
Add to cart
Type:
Burette Clamp (Single)
Price (SGD)
108.00
EA
Add to cart
For use with burettes. Azlon™ Burette Clamps contain rubber grips to prevent slipping.
  • Spring loaded polypropylene clamps that are corrosion resistant
  • Allow clear view of graduations along the full length of the burette
  • Rubber grips prevent slipping
  • An interchangeable compression cone on the central boss provides a firm grip on rods 11mm to 14mm in diameter
Specifications
TypeBurette Clamp (Single)
MaterialPolypropylene
IncludesRubber grip, interchangeable compression cone
For Use With (Equipment)Burette