Biohazard Incinerator Disposal Carton
Biohazard Incinerator Disposal Carton
Actual product may vary
Fisherbrand
Biohazard Incinerator Disposal Carton
Includes polyethylene liner. Bel Art™ Biohazard Incinerator Disposal Cartons are clearly marked for proper handling and processing.
Have Questions?
Catalog NumberCapacity (English)
FKS1132220414.4 gal.
Catalog number FKS11322204
Price (SGD)
455.00
PK
Add to cart
Capacity (English):
14.4 gal.
Price (SGD)
455.00
PK
Add to cart
  • Corrugated cardboard carton features fitted lid with flap
  • Lined with clear 1.5 mil polyethylene biohazard bag that resists tears, punctures
  • Bright biohazard markings for immediate and continuous identification
  • Instructions in English, Spanish, French and German
  • Use with Bel-Art reusable cover for biohazard carton (Mfr. No. F132040000)
  • Dimensions 12 x 12 x 27 in.
Specifications
Capacity (English)14.4 gal.
ColorRed, White
Dimensions (L x W x H)12 x 12 x 27 in. (30.5 x 30.5 x 68.6 cm)
Height (Metric)68.6 cm
Length (Metric)30.5 cm
Width (Metric)30.5 cm
AutoclavableNot Autoclavable
Liner MaterialPolyethylene
For Use With (Application)Proper handling and disposal of biohazard waste
Width (English)12 in.
Length (English)12 in.
Thickness1.5 mm
Height (English)27 in.
MaterialCardboard
Product TypeBiohazard Incinerator Carton Container
Unit SizePK