Whatman™ 1 Chr Chromatography Paper
Whatman™ 1 Chr Chromatography Paper
Actual product may vary
Fisherbrand
Whatman™ 1 Chr Chromatography Paper
Ensures the wicking capability and uniformity of capillary action important in chemical separations. Whatman™ 1 Chr Chromatography Paper is also widely used in protein and nucleic acid blotting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)Format
FKS11320724100 m2 cmRoll
FIS05-716-2S20 cm20 cmSheet
Catalog number FKS11320724
Price (SGD)
173.00
EA
Add to cart
Length (Metric):
100 m
Width (Metric):
2 cm
Format:
Roll
Price (SGD)
173.00
EA
Add to cart
Ensures the wicking capability and uniformity of capillary action important in chemical separations. Whatman™ 1 Chr Chromatography Paper is also widely used in protein and nucleic acid blotting.
Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind. Manufactured and tested specifically for chromatographic techniques to ensure the wicking capability and uniformity of capillary action important in chemical separations.
  • Typical Thickness: 0.18mm
  • Linear flowrate: 130mm/30 min.
  • Smooth surface
  • For general analytical separations
Specifications
Description1 Chr cellulose chromatography roll is a 0.18 mm paper for general analytical separations.
Dimensions (L x W)2.0 cm x 100 m (0.7 in. x 328.0 ft.)
Film Thickness0.18 mm
FinishSmooth
For Use With (Application)For General Analytical Separations
FormatRoll
GradeGrade 1 Chr
MaterialCellulose
Width (English)0.78 in.
Width (Metric)2 cm
Flow Rate130 mm/30 min.
Length (Metric)100 m
Unit SizeEA