Sterilin™ Quickstart Universal Polystyrene 30mL Containers
Sterilin™ Quickstart Universal Polystyrene 30mL Containers
Actual product may vary
Sterilin™ Quickstart Universal Polystyrene 30mL Containers
Sterilin™ Quickstart Universal Polystyrene 30mL Containers
Sterilin™ Quickstart Universal Polystyrene 30mL Containers
Fisherbrand
Sterilin™ Quickstart Universal Polystyrene 30mL Containers
Secure excellent temperature and chemical resistance with these 95kPa compliant 30mL leak-resistant transport containers.
Have Questions?
Catalog NumberLabelIncludes
FKS11319153PlainContainer Only
Catalog number FKS11319153
Price (SGD)
532.00
PK
Add to cart
Label:
Plain
Includes:
Container Only
Price (SGD)
532.00
PK
Add to cart
Secure excellent temperature and chemical resistance with these 95kPa compliant 30mL leak-resistant Thermo Scientific™ Sterilin™ Quickstart Universal Polystyrene 30mL Containers. Improved performance; easy to handle.
NOT AVAILABLE IN AMERICA
Features:
  • 1/2-turn cap ensures improved end user handling
  • Available pre-filled with boric acid (0.4g)
  • Supplied in convenient shelf packs of 50
  • Suitable for centrifugation at 3,800xg
  • Lot number on each container aids full traceability
Ordering Alerts:
Not available in all geographic regions
Specifications
Product TypeUniversal Container
MaterialPolystyrene
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)30 mL
Closure MaterialPolypropylene
DescriptionQuickstart Universal Container
Diameter (Metric)31 mm (cap), 24.5 mm (container)
Height (Metric)94 mm
IncludesContainer Only
LabelPlain
No. per Case400/Cs.
Unit SizePK