Sterilin™ Non-Pyrogenic Polystyrene Containers
Sterilin™ Non-Pyrogenic Polystyrene Containers
Actual product may vary
Fisherbrand
Sterilin™ Non-Pyrogenic Polystyrene Containers
Perform sterility and endotoxin tests with Thermo Scientific™ Sterilin™ Non-Pyrogenic Polystyrene Containers.
Have Questions?
Catalog NumberCapacity (Metric)Closure MaterialNo. per Case
FKS1131912330 mLPolypropylene400/Cs.
Catalog number FKS11319123
Price (SGD)
1,713.00
PK
Add to cart
Capacity (Metric):
30 mL
Closure Material:
Polypropylene
No. per Case:
400/Cs.
Price (SGD)
1,713.00
PK
Add to cart
Perform sterility and endotoxin tests with Thermo Scientific™ Sterilin™ Non-Pyrogenic Polystyrene Containers.
NOT AVAILABLE IN AMERICA
  • Sterilized by gamma irradiation
  • Leak-tested in accordance with BS EN ISO 6717
  • Labeled with lot numbers for full traceability
Specifications
Product TypeContainer
Closure IncludedYes
MaterialPolystyrene
ShapeCylindrical
SterilitySterile
Capacity (Metric)30 mL
Certifications/ComplianceLeak-tested in accordance with BS EN ISO 6717. Certified endotoxin free to levels <0.01EU/m.
Closure MaterialPolypropylene
DescriptionNon-Pyrogenic Container
Diameter (Metric)31 mm
Height (Metric)94 mm
LabelPrinted
No. per Case400/Cs.
Unit SizePK