Macrovolume Pipet Tips, For Gilson
Macrovolume Pipet Tips, For Gilson
Actual product may vary
Fisherbrand
Macrovolume Pipet Tips, For Gilson
For use with Gilson pipettes
Have Questions?
Catalog NumberVolume (Metric)
FKS113160941000 to 5000 μL
Catalog number FKS11316094
Price (SGD)
1,324.00
PK
Add to cart
Volume (Metric):
1000 to 5000 μL
Price (SGD)
1,324.00
PK
Add to cart
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are RNase-/DNase-free and nonpyrogenic. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Compatibility: Gilson P5000 and MLA pipettors
  • Graduation marks at 1, 2, 3, 4, 5 mL and 0.5 mL increments in between
Specifications
Volume (Metric)1000 to 5000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
AutoclavableAutoclavable
Clean ClaimsDNase, RNase, and Pyrogen free
FormatBulk
Tip StyleMacro
Packaging250 Tips/Pk, 10 Packs/Cs
For Use With (Equipment)MLA™ Pipette, Gilson™ Pipetman™ P10ML, Gilson™ Pipetman™ P5000
GraduatedYes
MaterialPolypropylene
Graduations1, 2, 3, 4, 5 mL and 0.5 mL Increments in Between
Unit SizePK