Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle
Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle
Actual product may vary
Fisherbrand
Whatman™ 602H 1/2 Qualitative Folded Filter Paper Circle
Dense filter paper forr collecting and removing fine precipitates
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS11315254240 mm
FKS11305254185 mm
Catalog number FKS11315254
Price (SGD)
983.00
PK
Add to cart
Diameter (Metric):
240 mm
Price (SGD)
983.00
PK
Add to cart
  • Used in sample preparation
  • Residual sugar determination
  • Acidic spectra
  • Refractometric analysis
  • Slow flow speed
  • HPLC
Specifications
DescriptionGrade 602 h ½ Qualitative Filter Paper Folded (Prepleated), 240 mm
FormatDisc
MaterialCellulose
Particle Retention<2 μm
Filtration SpeedApprox. 375 s
For Use With (Application)A dense prepleated filter paper for collecting very small particles and removing fine precipitates.
GradeGrade 602 h ½
Thickness (Metric)160 μm
Diameter (Metric)240 mm
Unit SizePK