Whatman™ Nylon Membranes
Whatman™ Nylon Membranes
Actual product may vary
Fisherbrand
Whatman™ Nylon Membranes
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)
FKS113148740.45 μm47 mm
FKS100121610.2 μm47 mm
FIS09-902-100.45 μm47 mm
Catalog number FKS11314874
Price (SGD)
982.00
PK
Add to cart
Pore Size:
0.45 μm
Diameter (Metric):
47 mm
Price (SGD)
982.00
PK
Add to cart
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
  • Pure nylon membranes
  • Tear-resistant
  • Filter many organic solvents
  • Autoclavable up to 135°C
  • Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
  • Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing
Specifications
AutoclavableAutoclavable
Filter TypeMembrane Filter
MaterialNylon
Pore Size0.45 μm
HydrophobicityHydrophilic
Quantity100/Pk.
TypeFiltration Membrane Disc
Diameter (Metric)47 mm
Unit SizePK