Store and transport 25 slides safely with durable ABS cases. Colored ABS 25 Place Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage.
Have Questions?
Change view
Catalog Number
Color
Lining Material
FKS11314135
Blue
Cork
FIS03-448-6
Blue
Foam
FIS03-448-8
Red
Foam
FIS03-448-10
White
Foam
FIS03-448-7
Green
Foam
FIS03-448-9
Yellow
Foam
6 Options
Catalog number FKS11314135
Price (SGD)
34.00
EA
Add to cart
Color:
Blue
Lining Material:
Cork
Price (SGD)
34.00
EA
Add to cart
Store and transport 25 slides safely with durable ABS cases. Fisherbrand™ Colored ABS 25 Place Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage.
Molded, rigid boxes securely hold slides in grooved slots
Both the lining and the slots are numbered to correspond to the slide inventory sheet
Cork or foam lining available
Friction-fit cover closes easily and allows for easy access to slides
Stackable
Specifications
TypeSlide Storage Box
Slide Capacity25
Closure TypeFriction-Fit Lid
ColorBlue
Device Holds25 Microscope Slides
Dimensions (L x W x H)141 x 88 x 35 mm
For Use With (Equipment)Standard 25 x 75 mm or 1 x 3 in. microscope slides