Benchtop Single Dispensing Bin
Benchtop Single Dispensing Bin
Actual product may vary
Fisherbrand
Benchtop Single Dispensing Bin
Access your lab materials with ease, even with gloved hands. Benchtop Single Dispensing Bin is ergonomically designed with a large, angled opening and an oversized lip.
Have Questions?
Catalog NumberColorDimensions (L x W x H)
FKS11314065Neon Blue155 x 155 x 170 mm
Catalog number FKS11314065
Price (SGD)
372.00
EA
Add to cart
Color:
Neon Blue
Dimensions (L x W x H):
155 x 155 x 170 mm
Price (SGD)
372.00
EA
Add to cart
Easily access your lab materials, even with gloved hands. Ideal for holding many different laboratory items including micro-centrifuge tubes.
Ergonomically designed:
  • Large, angled openings.
  • Oversized lip.
  • Lid remains open when tilted back.
Ideal for benchtop and wall mounted use:
  • Nonskid rubber feet.
  • Pre-drilled holes and included screws for wall mounting.
Specifications
TypeDispensing Bin
MaterialPMMA
ColorNeon Blue
ShapeSemi-circular
Quantity1 Ea.
Dimensions (L x W x H)155 x 155 x 170 mm
Length (English)6.1 in.
Length (Metric)155 mm
Width (Metric)155 mm
Height (Metric)170 mm
Unit SizeEA