Rubber Bulb
Rubber Bulb
Actual product may vary
Fisherbrand
Rubber Bulb
Rubber Bulb for use with large-volume pipets with 6 to 8 mm O.D.
Have Questions?
Catalog number FKS11314055
Price (SGD)
45.00
EA
Add to cart
Price (SGD)
45.00
EA
Add to cart
  • Constructed of Natural Rubber
  • 150 mL capacity, 103 mm L x 73 mm OD (6 mm hole dia.)
  • Reinforced ribs for use with large pipettes
Specifications
Compatible Tips6.35 mm
AutoclavableNot Autoclavable
ColorRed
MaterialRubber
ModelBulb
Number of Channels1
TypeBulb
Volume (Metric)150 mL
For Use With (Equipment)Large-volume pipets with 6 to 8 mm O.D.
Length (English)3.87 in.
Length (Metric)98 mm
Diameter (English) Opening0.25 in.
Diameter (English) Outer2.87 in.
Diameter (Metric) Opening6.35 mm
Diameter (Metric) Outer73 mm
Unit SizeEA