Blaubrand™ Class A Borosillicate Glass Tall Form Measuring Cylinder
Blaubrand™ Class A Borosillicate Glass Tall Form Measuring Cylinder
Actual product may vary
Fisherbrand
Blaubrand™ Class A Borosillicate Glass Tall Form Measuring Cylinder
Manufactured from Borosilicate Glass and complies with DIN ISO 4788 standards. BRAND™ Blaubrand™ Class A Borosilicate Glass Tall Form Measuring Cylinder includes integral spout and hexagonal glass base for stability. Graduations are marked in blue enamel for easy volume assessment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS11313894250 mL2 mL
FKS11303894100 mL1 mL
Catalog number FKS11313894
Price (SGD)
192.00
PK
Add to cart
Capacity (Metric):
250 mL
Graduations:
2 mL
Price (SGD)
192.00
PK
Add to cart
  • Manufactured from Borosilicate Glass
  • Complies with DIN ISO 4788 standards
  • Calibrated to contain (TC, In.)
  • Includes integral spout and hexagonal glass base for stability
  • Graduations marked in blue enamel for easy volume assessment
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorBlue
ShapeRound
Certifications/ComplianceDIN EN ISO 4788
ClassA
Height (Metric)335 mm
Tolerance Metric±1 mL
TypeGraduated Measuring Cylinder
MaterialBorosilicate Glass
Capacity (Metric)250 mL
IncludesBatch Certificate
ScaleDE-M marking, blue enamel graduations
Graduations2 mL
AutoclavableAutoclavable
Subdivision2mL
Unit SizePK