Fast-Release Pipette Pump™ II Pipetters
Fast-Release Pipette Pump™ II Pipetters
Actual product may vary
Fisherbrand
Fast-Release Pipette Pump™ II Pipetters
Includes a rapid release valve for quick dispensing. Pipetters are color coded by size for easy identification.
Have Questions?
Catalog NumberColorFor Use With (Application)
FKS11312244GreenPipets up to 10mL
Catalog number FKS11312244
Price (SGD)
160.00
EA
Add to cart
Color:
Green
For Use With (Application):
Pipets up to 10mL
Price (SGD)
160.00
EA
Add to cart
  • Rotate the knurled thumb wheel to aspirate liquid to the desired volume
  • Apply light pressure to the fast release lever to rapidly dispense
  • Rotate the thumb wheel for gradual dispensing
  • Glass or plastic pipettes are easily inserted into a unique collar with stabilizing fingers for a secure hold
  • Sizes are color coded; can be disassembled for cleaning
Specifications
Adjustable Tip SpacingNo
AutoclavableNot Autoclavable
ColorGreen
DisposableReusable
For Use WithSerologic Pipette
Grip TypeErgonomic
MaterialPolypropylene, Silicone Rubber
ModelPipette Pump II
Number of Channels1
SterilityNon-sterile
TypeFast Release Pipet Filler
Volume (Metric)10 mL
For Use With (Application)Pipets up to 10mL
For Use With (Equipment)10mL
Unit SizeEA