Whatman™ Quantitative Filter Paper: Grade 41 Sheets
Whatman™ Quantitative Filter Paper: Grade 41 Sheets
Actual product may vary
Whatman™ Quantitative Filter Paper: Grade 41 Sheets
Use for quantitative air pollution analysis as a paper tape for impregnation when determining gaseous compounds at high flow rates or for quantitative air pollution analysis. Fisherbrand™ Quantitative Filter Paper: Grade 41 Sheets are commonly used in preparing samples for metal analysis after nitric acid digestion.
Have Questions?
Catalog NumberDimensions (L x W)
FKS11310474570 x 460 mm
Catalog number FKS11310474
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
570 x 460 mm
Use for quantitative air pollution analysis as a paper tape for impregnation when determining gaseous compounds at high flow rates or for quantitative air pollution analysis. Fisherbrand™ Quantitative Filter Paper: Grade 41 Sheets are commonly used in preparing samples for metal analysis after nitric acid digestion.
Ashless grades are very pure filters suitable for a wide range of critical analytical filtration procedures. Whatman™ quantitative filter papers are designed for gravimetric analysis and the preparation of samples for instrumental analysis.
  • Extremely low ash content (0.007% ash maximum)
  • 20μm nominal particle retention rating
  • Fast filtration speed (Herzberg 54 s)
  • Commonly used for routine air pollution analysis and soil pollution analysis
  • High-purity filters for quantitative analysis
Specifications
Airflow Rate4 s/100 mL/in2
DescriptionGrade 41 Fast Ashless Filter Paper,460 × 570 mm sheet (100 pcs)
Filter TypeAshless
FormatSheet
MaterialCotton Linters
Particle Retention20 μm
Pore Size20 μm
Filtration SpeedASTM, 12 seconds; Herzberg, 54 seconds
Flow RateFast
For Use With (Application)Designed for gravimetric analysis and the preparation of samples for instrumental analysis.For analytical procedures involving coarse particles or gelatinous precipitates (e.g., iron or aluminum hydroxides).Also used in quantitative air pollution analys
GradeGrade 41
PorosityCoarse
Quantity100 Pk.
Thickness (Metric)215 μm
TypeQuantitative Filter Paper
Dimensions (L x W)570 x 460 mm
Length (Metric)570 mm
Width (Metric)460 mm
Unit SizePK