Innova™ 43R Incubator Shaker
Innova™ 43R Incubator Shaker
Actual product may vary
Fisherbrand
Innova™ 43R Incubator Shaker
Use these versatile incubator shakers for support high-speed applications and heavy workloads. New Brunswick Scientific™ Innova™ 43R Incubator Shakers handle large quantities or large volumes of cultures with precision and ease.
Have Questions?
Catalog NumberOrbit
FKS1131002525 mm
Catalog number FKS11310025
Price (SGD)
102,842.00
EA
Add to cart
Orbit:
25 mm
Price (SGD)
102,842.00
EA
Add to cart
  • Console style shaker/incubator with triple-eccentric drive – provides large capacity shaking for flasks up to 6 liters
  • Programmable Innova controls – automatically changes temperature, speed, and optional photosynthetic and UV germicidal lights at timed intervals
  • Top mounted controls and display – for easy, ergonomic viewing and setting
  • Foot pedal release with easy opening lid – provides hands free operation
  • RS-232 port for data logging
  • Built-in water reservoir humidifies chamber to reduce sample evaporation, while also protecting unit from spills and including a drain for easy cleaning
Specifications
Orbit25 mm
Speed Range25 to 500 rpm
Electrical Requirements230 V, 50 Hz
Voltage230 V
Holds6 L
Product TypeIncubator Shaker
Frequency50 Hz
Depth (Metric)81.3 cm
Height (Metric)165.1 cm
Width (Metric)115.6 cm
ModelInnova 43R
Timer Range0.01 to 99:59 hr.
Unit SizeEA