Search Thermo Fisher Scientific
| Catalog Number | Height (Metric) | No. per Case | Sterility | Vent |
|---|---|---|---|---|
| FKS11309283 | 16.1 mm | 500 | Non-sterile | 3 |
| FKS11389273 | 15.9 mm | 500 | Non-sterile | 1 |
| FKS11759252 | 16.1 mm | 500 | Sterile | 3 |
| FKS11739252 | 15.7 mm | 500 | Sterile | No |
| FKS15370366 | 15.7 mm | 500 | Non-sterile | No |
Culture microorganisms on solid media using Thermo Scientific™ Sterilin™ Standard 90mm Petri Dishes. These shallow dishes maximize incubator space and feature a mirror finish that ensures high optical quality.
NOT AVAILABLE IN AMERICA
Features:
Available Vent Formats: