Glass-Disposal Boxes
Actual product may vary
Glass-Disposal Boxes
Glass-Disposal Boxes
Glass-Disposal Boxes
Fisherbrand
Glass-Disposal Boxes
Sturdy glass disposal box manufactured from corrugated cardboard supplied with polyethylene liner bags. Fisherbrand™ Glass-Disposal Boxes keep glass waste separate from other waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelDimensions (L x W x H)
FKS11308503Floor12 x 12 x 27 in.
FIS12-009-7BBenchtop8 x 8 x 10 in.
FKS11318503Benchtop8 x 8 x 10 in.
FIS17-988-448Intermediate12 x 12 x 20 in.
FKS11507692Intermediate12 x 12 x 20 in.
FIS12-009-7AFloor12 x 12 x 27 in.
Catalog number FKS11308503
Price (SGD)
132.00
Pack
Model:
Floor
Dimensions (L x W x H):
12 x 12 x 27 in.
Price (SGD)
132.00
Pack
Sturdy glass disposal box manufactured from polyethylene-lined corrugated cardboard. Fisherbrand™ Glass-Disposal Box, Benchtop keeps glass waste separated from other waste.
  • Floor model glass disposal box with handles is safer in use and disposal
  • Lid covers the built-in integral handles when use is not necessary
  • Sturdy, corrugated cardboard
  • Tough polyethylene liner
  • Identifying graphics with instructions
  • Tight-fitting lid has flap opening to accept material
  • Folding design for shipping and storage
  • Available in a range of convenient sizes
Specifications
Closure TypeIntegral Safety Lid
Dimensions (L x W x H)12 x 12 x 27 in.
Height (Metric)69 cm
Length (Metric)30 cm
ModelFloor
Quantity6/Cs.
Width (Metric)30 cm
Liner MaterialPolyethylene
Width (English)12 in.
Length (English)12 in.
Height (English)27 in.
MaterialCorrugated Cardboard
Unit SizePack