Whatman™ GD/X PVDF Syringe Filters
Whatman™ GD/X PVDF Syringe Filters
Actual product may vary
Fisher Catalogue
Whatman™ GD/X PVDF Syringe Filters
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X PVDF Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)Quantity
FKS113047740.45 μm25 mm150/Pk.
FIS09-927-28D0.45 μm25 mm150/Pk.
Catalog number FKS11304774
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.45 μm
Diameter (Metric):
25 mm
Quantity:
150/Pk.
Processes three to seven times more sample volume than filters without prefilter. Whatman™ GD/X PVDF Syringe Filters are specifically designed for filtration of viscous or otherwise hard-to-filter samples with high solids content.
Constructed of a polypropylene housing with a prefiltration stack of Whatman GMF 150 (graded density) and GF/F glass microfiber prefilters. This design prevents the buildup of back pressure typically caused by the blocking of an unprotected membrane.
  • Syringe filter with integrated prefiltration
  • Female Luer-Lok™ inlet; male slip Luer outlet
  • Flow should enter from the inlet
  • Autoclavable at 121°C (250°F) at 15psi for 20 min.
  • Maximum temperature, 131°C (267°F) Filtration area: 13mm - 1.3cm2; 25mm - 4.6cm2 Pressure limit: 13mm - 100psi; 25mm - 75psi
  • All materials biosafe—pass USP Class VI requirements
  • Prefiltration media: 100% Borosilicate glass fiber GMF 150 10μm: 1μm GF/F 0.7μm
  • Disposable filtration devices provide labor saving efficiency while ensuring consistent filter performance
  • High loading capacity for samples with high solids content
  • Suitable for capillary electrophoresis, HPLC, and UV/VIS sample preparation
Nonsterile GD/X filters are available in two diameters: 13 mm and 25 mm, and three pore sizes: 0.2 μm, 0.45 μm, and 5 μm
Specifications
DisposableSingle-use
Filter TypeSyringe Filter
Material (Membrane)PVDF
Pore Size0.45 μm
Filtration Area4.6 cm2
Housing MaterialPigment free polypropylene (PP)
Inlet StyleFemale Luer Lock
Outlet StyleMale Luer
Prefilter100% Borosilicate Glass FiberGMF 150 10 μm: 1 μmGF/F 0.7 μm
Pressure Rating5.2 bar (75 psi)
Quantity150/Pk.
Volume (Metric)≤10 mL
Dimensions (L x W x H)21.6 x 29.8 mm
Diameter (Metric)25 mm
Unit SizePK