Finned Plug for Disposable Tubes
Finned Plug for Disposable Tubes
Actual product may vary
Fisherbrand
Finned Plug for Disposable Tubes
Designed with flexible fins to create a snug fit.
Have Questions?
Catalog NumberColor
FKS11301205Red
Catalog number FKS11301205
Price (SGD)
81.00
PK
Add to cart
Color:
Red
Price (SGD)
81.00
PK
Add to cart
Designed with flexible fins to create a snug fit.
  • Fits tubes 11 to 13mm or 15 to 17mm 
  • Available in a variety of colors for coding
Specifications
Certifications/ComplianceCE-IVD
Product TypeFinned Plug
MaterialPolyethylene
ColorRed
For Use With (Equipment)11 to 13 mm Tubes
Unit SizePK