Whatman™ Benchkote Surface Protector Sheets
Whatman™ Benchkote Surface Protector Sheets
Actual product may vary
Fisherbrand
Whatman™ Benchkote Surface Protector Sheets
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FKS1130067450 Sheets
FIS09-800-976100 Sheets
Catalog number FKS11300674
Price (SGD)
611.00
PK
Add to cart
Quantity:
50 Sheets
Price (SGD)
611.00
PK
Add to cart
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
  • Strong: Benchkote surface protector is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
  • Dimensions: 460 x 570mm
Specifications
Product TypeSurface Protector
MaterialPolyethylene
Absorbancy750 mL/m2
Length (English)18 in.
Length (Metric)460 mm
Width (English)22.5 in.
Width (Metric)570 mm
Dimensions (L x W)460 x 570mm
For Use With (Application)Suitable for saturation with disinfectant
Quantity50 Sheets
Unit SizePK