Tubing Connector for Use with Peristaltic Pump P-1
Tubing Connector for Use with Peristaltic Pump P-1
Actual product may vary
Fisherbrand
Tubing Connector for Use with Peristaltic Pump P-1
Used for the connection of 1.8mmOD/1.5mmID capillary tubing with the Peristatic Pump P-1.
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS113000821.5 mm
Catalog number FKS11300082
Price (SGD)
851.00
PK
Add to cart
Diameter (Metric) Inner:
1.5 mm
Price (SGD)
851.00
PK
Add to cart
Instructions for the connection of capillary tubing:
  1. Cut the tubing cleanly at a 45° angle
  2. Remove the screw nut and slip it over the tubing
  3. Remove the sealing plug
  4. Push the tubing through the sealing plug until it protrudes about 1cm
  5. Seat the tubing in the nipple: slide the sealing plug along the tubing and into the nipple
  6. Finger-tighten the connector
Specifications
TypeTubing Connector
Diameter (Metric) Outer1.8 mm
ConnectionsBarb-stepped
MaterialPolypropylene, POM Fluoro Rubber
Diameter (Metric) Inner1.5 mm
For Use With (Equipment)Peristaltic Pump P-1
Unit SizePK