Heratherm™ Advanced Protocol Microbiological Incubator, 178 L, Stainless Steel Exterior
Product Image
Fisherbrand
Heratherm™ Advanced Protocol Microbiological Incubator, 178 L, Stainless Steel Exterior
Temperature Range: Ambient +5°C to 105°C
Have Questions?
Catalog NumberVoltage
FKS10859970230 V
Catalog number FKS10859970
Price (SGD)
27,627.00
EA
Add to cart
Voltage:
230 V
Price (SGD)
27,627.00
EA
Add to cart
Featuring outstanding temperature performance with innovative dual convection technology, Thermo Scientific™ Heratherm Advance Protocol Microbiological Incubators provide an optimal sample environment—the flexible tool in the lab to be used for multiple applications. Choice of stainless steel exterior available for demanding pharmaceutical and clinical requirements.
  • Dual convection for application versatility—fan speed adjustable from 0 to 100%
  • Advanced digital timer for daily or weekly ON / OFF cycles
  • Easy to clean, corrosion-resistant stainless-steel interior (AISI 304)
  • Broad temperature range from 5°C above ambient to 105°C—suitable for drying applications
  • Temperature uniformity of ± 0.6 (Fan off) and between ±0.2° and ± 0.4C (Fan full speed) (measured at 37°C)
  • Temperature stability of ±0.1°C (measured at 37°C)
  • Intuitive user interface for setting temperature
  • Large, easy to read vacuum fluorescent display
  • Internal glass door allows sample viewing without impacting temperature
  • Exceptionally small footprint
Ordering Information:
Availability may vary by country.
Specifications
Capacity (English)6.3 cu. ft.
Capacity (English) Shelf55 lb.
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)178 L
Convection TechnologyDual convection
MaterialStainless Steel
No. of Shelves2 std./19 max.
Plug TypePlug type provided based on country
Temperature Range (Metric)Ambient +5° to 105°C
TypeMicrobiological Incubator
Temperature Stability±0.1°C
Temperature Uniformity±0.6°C
Timerweekly / real time / hour
Dimensions (D x W x H) Interior21.4 x 18.3 x 27.9 in. (54.3 x 46.4 x 70.8 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Length (English)29.1 in.
Length (Metric)73.8 cm
Width (English)25.2 in.
Width (English) Interior18.3 in.
Width (Metric)64 cm
Width (Metric) Interior46.4 cm
Height (English)36.2 in.
Height (English) Interior27.9 in.
Height (Metric)92 cm
Height (Metric) Interior70.8 cm
Depth (English) Interior21.4 in.
Depth (Metric) Interior54.3 cm
Weight (English)154 lb.
Weight (Metric)70 kg
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Wattage1300 w
Amperage5.7 A