Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) OuterSocket Joint
FKS10840861250 mL130 mm85 mm24/29
FKS10508980100 mL100 mm64 mm24/29
Catalog number FKS10840861
Price (SGD)
531.00
PK
Add to cart
Capacity (Metric):
250 mL
Height (Metric):
130 mm
Diameter (Metric) Outer:
85 mm
Socket Joint:
24/29
Price (SGD)
531.00
PK
Add to cart
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Glass Erlenmeyer Flask with Quickfit Ground Glass Socket complies to ISO 4797.
  • Complies to ISO 4797
  • Conical flasks with highest quality Quickfit™ ground glass socket
  • Borosilicate glass for corrosion free performance and high resistance to thermal shock
Specifications
Certifications/ComplianceISO 4797
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Height (Metric)130 mm
IncludesGround socket
ShapeConical
Socket Joint24/29
Diameter (Metric) Outer85 mm
Unit SizePK