Pipetboy acu 2 Pipette Controller
Pipetboy acu 2 Pipette Controller
Actual product may vary
Fisher Catalogue
Pipetboy acu 2 Pipette Controller
Pipet with precision. Integra Biosciences™ Pipetboy acu 2 Pipette Controller represents the most popular pipet aid with the colorful look and the weight balance.
Have Questions?
Catalog NumberColor
FKS10798252Blue
Catalog number FKS10798252
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Blue
  • Cable free pipette controller.
  • Extremely lightweight, compact and easy to use.
  • Works on a rechargeable 9V NiMH battery which is easily inserted at the top of the handle.
  • One charge is sufficient to fill and purge at least 500 x 50mL pipettes.
  • Accepts standard pipettes ranging from 0.1mL to 100mL.
  • Comes in a range of five transparent colors and a traditional light grey to aid identification.
  • With battery, transformer for 230V/50Hz, sterile filter and wall mount.
Specifications
Battery TypeNickel-metal hydride
ColorBlue
Pore Size0.45 μm
TypePipette Holder
Filter TypeHydrophobic membrane
Voltage230 V
Frequency50 Hz
Unit SizePK