Used as a certified conductivity standards. Reagecon™ Conductivity Standards NIST Traceable, 100000μS/cm is traceable to NIST reference materials.
Have Questions?
Catalog Number
Quantity
FKS10790320
500 mL
Catalog number FKS10790320
Price (SGD)
565.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
565.00
EA
Add to cart
Specifications
Quantity500 mL
Unit SizeEA
Accurate to ±1%
Traceable to NIST
GLP compliant
Certificate of analysis supplied with each bottle (including method of analysis and standard reference material detail)
Wide range: 20 values from 1.3μS/cm to 500,000μS/cm inclusive
An enhanced shelf life is assured through the use of proprietary technologyAccurate conductivity measurement requires periodic calibration of the conductivity sensor in order to determine its cell constant (the multiplier used to convert the sensor's output to the displayed conductivity). Our standard value conductivity standards match the pre-programmed values used by all leading brands of conductivity meters for automatic calibration, While the broad range standards allow flexibility.