Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Blue Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
Have Questions?
Catalog Number
Volume (Metric)
FKS10787524
200 to 1000 μL
Catalog number FKS10787524
Price (SGD)
32.00
PK
Add to cart
Volume (Metric):
200 to 1000 μL
Price (SGD)
32.00
PK
Add to cart
Made of high quality polypropylene
Tight fit due to the precisely fitting cone
Optimal handling due to the stable shape
Autoclavable
Guaranteed dust-free due to special ionization treatment
Available pre-sterilized or non-sterile
Excellent transparency due to high quality surfaces
Minimal residual wetting due to controlled quality of raw materials
Excellent volume accuracy due to precise tip orifice
Practice-oriented tip volumes
Aeroject™ tips with special filters made of hydrophobic PE-HD as
Aerosol barrier and for protection against contamination