ZM 200 Model Ultra-Centrifugal Mill
ZM 200 Model Ultra-Centrifugal Mill
Actual product may vary
Fisherbrand
ZM 200 Model Ultra-Centrifugal Mill
Ensure the gentle preparation of analytical samples in a very short time. Retsch™ ZM 200 Model Ultra-Centrifugal Mills are versatile instruments that can be easily adapted to varying applications.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS10786671515 x 410 x 365 mm
Catalog number FKS10786671
Price (SGD)
42,333.00
EA
Add to cart
Dimensions (L x W x H):
515 x 410 x 365 mm
Price (SGD)
42,333.00
EA
Add to cart
  • Gentle and very rapid size reduction by pre and fine grinding in one run
  • Powerdrive with optimally matched frequency converter and 3 phase motor
  • Comfortable parameter setting via display and ergonomic 1 button operation
  • Wide range of accessories including various collection and feeding systems, rotors and sieves
  • Wide speed range
  • Patented cassette system for maximum sample recovery and easy cleaning
  • Easily exchangeable grinding and sieve inserts
  • Defined final fineness due to ring sieves with aperture sizes from 0.08mm to 10mm
  • Comfortable safety housing with automatic cover closure
  • Motor compartment and electronics protected against dust and material penetration
  • Ideal for fine grinding for sample preparation of activated charcoal, animal tissue, bentonite, bones, cereals, chemical products, coal, coffee beans, coke, collagen, corn, dried fruit and vegetables, dried larvae, drugs, electronic components, spices and straw etc.
Specifications
Product TypeUltra-Centrifugal Mill
Certifications/ComplianceCE Certified
Dimensions (L x W x H)515 x 410 x 365 mm
For Use With (Application)For Fine Grinding
Height (Metric)365 mm
Hertz50/60 Hz
Length (Metric)515 mm
ModelZM 200
Watts1300 W
Width (Metric)410 mm
Unit SizeEA